Anti-CD79b Armenian Hamster Monoclonal Antibody [clone: HM79-12]

Supplier: BioLegend
132801 132802 132802 132801
132801-BLEA 127.38 USD
CA132802-BL CA132801-BL 132801-BL 132802-BL
Anti-CD79b Armenian Hamster Monoclonal Antibody [clone: HM79-12]
Antibodies
Purified anti-mouse CD79b (Igβ) [HM79-12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD79b
Clonality: Monoclonal
Clone: HM79-12
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR