Anti-CD317 (BST2, PDCA-1) Rat Monoclonal Antibody [clone: 927]

Supplier: BioLegend
127002 127002
127002-BLEA 158.69 USD
CA127002-BL 127002-BL
Anti-CD317 (BST2, PDCA-1) Rat Monoclonal Antibody [clone: 927]
Antibodies
Purified anti-mouse CD317 (BST2, PDCA-1) [927]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IF, IP; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD317 (BST2, PDCA-1)
Clonality: Monoclonal
Clone: 927
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR