Anti-EBI3 Mouse Monoclonal Antibody [clone: V1.4F5.25]

Supplier: Rockland Immunochemicals
210-301-C56
RL210-301-C56EA 528.35 USD
RL210-301-C56
Anti-EBI3 Mouse Monoclonal Antibody [clone: V1.4F5.25]
Antibodies
Type: Primary
Antigen: EBI3
Clonality: Monoclonal
Clone: V1.4F5.25
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR