Anti-CD45.1 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: A20]

Supplier: BioLegend
110733 110733
110733-BLEA 225.83 USD
CA110733-BL 110733-BL
Anti-CD45.1 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: A20]
Antibodies
Brilliant Violet 570™ anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 125 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: A20
Conjugation: Brilliant Violet® 570
Epitope:
Host: Mouse
Isotype: Mouse (A.SW) IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR