Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: RM4-5]

Supplier: BioLegend
100542 100542 100541 100541
10756-664EA 305.67 USD
CA10756-664 10756-664 100541-BL CA100541-BL
Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 570) [clone: RM4-5]
Antibodies
Brilliant Violet 570™ anti-mouse CD4 [RM4-5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunofluorescent staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RM4-5
Conjugation: Brilliant Violet® 570
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR