Anti-CD19 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 6D5]

Supplier: BioLegend
115538 115538 115549 115537 115537 115549
10756-924EA 332.84 USD
10756-924 CA10756-924 CA115538-BL CA115537-BL 115538-BL 115537-BL
Anti-CD19 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 6D5]
Antibodies
Brilliant Violet 421™ anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. For immunofluorescent staining using μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 6D5
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR