Anti-IL-10 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: JES3-9D7]

Supplier: BioLegend
501421 501421
501421-BLEA 205.32 USD
501421-BL CA501421-BL
Anti-IL-10 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: JES3-9D7]
Antibodies
Brilliant Violet 421™ anti-human IL-10 [JES3-9D7]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Rhesus, Cynomolgus; Apps: ICFC; Size: 25 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: JES3-9D7
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Human;Cross-Reactivity: Baboon;Rhesus;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR