Anti-SHB Rabbit Polyclonal Antibody

Supplier: Abcepta
AP20620A
10217-922EA 580.86 USD
10217-922
Anti-SHB Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:25, Flow Cytometry: 1:25,

Type: Primary
Antigen: SHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR