Anti-CD69 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: FN50]

Supplier: BioLegend
310929 310929 310930 310930
10758-026EA 514.34 USD
CA10758-026 10758-026 CA310929-BL 310929-BL
Anti-CD69 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: FN50]
Antibodies
Brilliant Violet 421™ anti-human CD69 [FN50]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR