Anti-CD5 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: UCHT2]

Supplier: BioLegend
300625 300625 300626 300626
300625-BLEA 196.67 USD
300625-BL 300626-BL CA300625-BL CA300626-BL
Anti-CD5 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: UCHT2]
Antibodies
Brilliant Violet 421™ anti-human CD5 [UCHT2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Capuchin Monkey, Common Marmoset, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Capuchin Monkey;Common Marmoset;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR