Anti-CD284 (TLR4) Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HTA125]

Supplier: BioLegend
312811 312811
312811-BLEA 307.11 USD
CA312811-BL 312811-BL
Anti-CD284 (TLR4) Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HTA125]
Antibodies
Brilliant Violet 421™ anti-human CD284 (TLR4) [HTA125]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Guinea Pig; Apps: FC; Size: 25 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD284 (TLR4)
Clonality: Monoclonal
Clone: HTA125
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Cross-Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR