Anti-CD1a Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HI149]

Supplier: BioLegend
300127 300128 300127
10020-372EA 422.75 USD
10020-372 300127-BL CA300127-BL
Anti-CD1a Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: HI149]
Antibodies
Brilliant Violet 421™ anti-human CD1a [HI149]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD1a
Clonality: Monoclonal
Clone: HI149
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR