Anti-IgM Mouse Monoclonal Antibody (Biotin) [clone: MRM-47]

Supplier: BioLegend
408903 408903
408903-BLEA 205.1 USD
CA408903-BL 408903-BL
Anti-IgM Mouse Monoclonal Antibody (Biotin) [clone: MRM-47]
Antibodies
Biotin anti-rat IgM [MRM-47]; Isotype: Mouse IgG; Reactivity: Rat; Apps: FC, ELISA, IHC-F; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. <span> For immunohistochemistry, a concentration range of 2.5 - 5 μg/ml is suggested. </span>It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgM
Clonality: Monoclonal
Clone: MRM-47
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR