Anti-TNF-a Rat Monoclonal Antibody (Biotin) [clone: MP6-XT22]

Supplier: BioLegend
506312 506312 506311 506311
506312-BLEA 291.43 USD
CA506311-BL CA506312-BL 506312-BL 506311-BL
Anti-TNF-a Rat Monoclonal Antibody (Biotin) [clone: MP6-XT22]
Antibodies
Biotin anti-mouse TNF-α [MP6-XT22]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Detection, ELISPOT Detection, ICFC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.25-1.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of mouse TNF-α recombinant protein ranging from 500 to 4 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TNF-a
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR