Anti-IL-1b Goat Polyclonal Antibody (Biotin) [clone: Poly5158]

Supplier: BioLegend
515801 515801
515801-BLEA 403.05 USD
CA515801-BL 515801-BL
Anti-IL-1b Goat Polyclonal Antibody (Biotin) [clone: Poly5158]
Antibodies
Biotin anti-mouse IL-1β [Poly5158]; Isotype: Goat Polyclonal Ig; Reactivity: Mouse; Apps: ELISA Detection; Size: 5 Plates

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection, the suggested use of this reagent is 1:200 dilution. To obtain a linear standard curve, serial dilutions of IL-1β recombinant protein ranging from 2000 to 30 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-1b
Clonality: Polyclonal
Clone: Poly5158
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat Polyclonal Ig
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR