Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL-12/IL-23 Rat Monoclonal Antibody (Biotin) [clone: C17.8]

Supplier: Biolegend
505301 505302 505301 505302
505301-BL CA505302-BL CA505301-BL 505302-BL
Anti-IL-12/IL-23 Rat Monoclonal Antibody (Biotin) [clone: C17.8]
Biotin anti-mouse IL-12/IL-23 p40 [C17.8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ELISA Detection, ELISPOT Detection, ICFC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.25-1.0 μg/ml is recommended. For use as an ELISPOT detection antibody, a concentration range of 0.5-2.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-12 recombinant protein ranging from 250 to 2 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-12/IL-23
Clonality: Monoclonal
Clone: C17.8
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;IL-12/IL-23 p40 subunit (monomer;homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40)

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty