Anti-CD194 (CCR4) Armenian Hamster Monoclonal Antibody (Biotin) [clone: 2G12]

Supplier: BioLegend
131215 131216 131215 131216
131215-BLEA 133.83 USD
131215-BL CA131216-BL CA131215-BL 131216-BL
Anti-CD194 (CCR4) Armenian Hamster Monoclonal Antibody (Biotin) [clone: 2G12]
Antibodies
Biotin anti-mouse CD194 (CCR4) [2G12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD194 (CCR4)
Clonality: Monoclonal
Clone: 2G12
Conjugation: Biotin
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR