Anti-CD140a Rat Monoclonal Antibody (Biotin) [clone: APA5]

Supplier: BioLegend
135909 135909 135910 135910
135909-BLEA 63.69 USD
CA135909-BL CA135910-BL 135909-BL 135910-BL
Anti-CD140a Rat Monoclonal Antibody (Biotin) [clone: APA5]
Antibodies
Biotin anti-mouse CD140a [APA5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD140a
Clonality: Monoclonal
Clone: APA5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR