Anti-CD135 Rat Monoclonal Antibody (Biotin) [clone: A2F10]

Supplier: BioLegend
135307 135308 135308 135307
135307-BLEA 89.58 USD
135307-BL CA135308-BL CA135307-BL 135308-BL
Anti-CD135 Rat Monoclonal Antibody (Biotin) [clone: A2F10]
Antibodies
Biotin anti-mouse CD135 [A2F10]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD135
Clonality: Monoclonal
Clone: A2F10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR