Anti-IgG Goat Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20129 20129-1
89138-662EA 208.83 USD
89138-662 71004-186
Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 633
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR