Anti-IgG Donkey Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20124-1 20124
89138-644EA 309.56 USD
89138-644 89138-646
Anti-IgG Donkey Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 633
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR