Anti-IgG2a Goat Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20259 20259-1
71003-866EA 127.9 USD
71003-866 71003-864
Anti-IgG2a Goat Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR