Anti-IgG Chicken Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20221-1 20221
71003-884EA 208.83 USD
71003-884 71003-886
Anti-IgG Chicken Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgY
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR