Anti-IgG Donkey Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20159-1 20159
89138-754EA 103.9 USD
89138-754 89138-752
Anti-IgG Donkey Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR