Anti-IgG Goat Polyclonal Antibody (CF™ 568)

Supplier: Biotium

20097 20097-1
89138-564EA 103.9 USD
89138-564 89138-562
Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 568
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR