Anti-IgG Goat Polyclonal Antibody (CF™ 568)

Supplier: Biotium

20096 20096-1
89138-560EA 103.9 USD
89138-560 89138-558
Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 568
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR