Anti-IgG Donkey Polyclonal Antibody (CF™ 555)

Supplier: Biotium

20276 20276-1
71004-018EA 249.98 USD
71004-018 71004-020
Anti-IgG Donkey Polyclonal Antibody (CF™ 555)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 555
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR