Anti-IgG Goat Polyclonal Antibody (CF™ 555)

Supplier: Biotium

20231-1 20231
89138-786EA 103.9 USD
89138-786 89138-784
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 555
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR