Anti-SUMO1 Mouse Monoclonal Antibody

Supplier: Abcepta
AM1200B
80083-384EA 785.02 USD
80083-384
Anti-SUMO1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SUMO1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR