Anti-PTPN1 Mouse Monoclonal Antibody

Supplier: Abcepta
AM8411B
60941-140EA 789.68 USD
60941-140
Anti-PTPN1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PTPN1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR