Anti-STAT2 Rabbit Polyclonal Antibody [clone: Poly6245]

Supplier: BioLegend
624502 624502
624502-BLEA 316.31 USD
CA624502-BL 624502-BL
Anti-STAT2 Rabbit Polyclonal Antibody [clone: Poly6245]
Antibodies
Purified anti-STAT2 [Poly6245]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human; Apps: WB, IP; Size: 200 μl

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. For IHC, use a 1:50 dilution of antibody for staining. Antigen retrieval for IHC of formalin-fixed paraffin-embedded tissue using 0.01 M sodium citrate buffer is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: STAT2
Clonality: Polyclonal
Clone: Poly6245
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR