Anti-SSRP1 Mouse Monoclonal Antibody [clone: 3E4]

Supplier: BioLegend
609802 609802
609802-BLEA 235.31 USD
CA609802-BL 609802-BL
Anti-SSRP1 Mouse Monoclonal Antibody [clone: 3E4]
Antibodies
Purified anti-SSRP1 [3E4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IF, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg per 5 ml antibody dilution buffer for each mini-gel. For immunofluorescence microscopy, a concentration range of 2.5-10 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SSRP1
Clonality: Monoclonal
Clone: 3E4
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR