Anti-PTPN11 Mouse Monoclonal Antibody [clone: 246CT9.8.2]

Supplier: Abcepta
AM1920A
89512-248EA 606.77 USD
89512-248
Anti-PTPN11 Mouse Monoclonal Antibody [clone: 246CT9.8.2]
Antibodies
Western Blot: 1:100~500,

Type: Primary
Antigen: PTPN11
Clonality: Monoclonal
Clone: 246CT9.8.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR