Anti-Sp3 Rabbit Polyclonal Antibody [clone: Poly6248]

Supplier: BioLegend
624802 624801 624801 624802
624802-BLEA 286.85 USD
CA624801-BL 624802-BL CA624802-BL 624801-BL
Anti-Sp3 Rabbit Polyclonal Antibody [clone: Poly6248]
Purified anti-Sp3 [Poly6248]; Isotype: Rabbit Polyclonal IgG; Reactivity: Mouse, Rat, Human; Apps: WB, IHC; Size: 200 μl

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. For IHC, use a 1:50 dilution of antibody for staining. Antigen retrieval for IHC of formalin-fixed paraffin-embedded tissue using 0.01 M sodium citrate buffer is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Sp3
Clonality: Polyclonal
Clone: Poly6248
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR