Anti-GAD2 Mouse Monoclonal Antibody [clone: 458CT17.2.4]

Supplier: Abcepta
AM2049A
89512-110EA 637.11 USD
89512-110
Anti-GAD2 Mouse Monoclonal Antibody [clone: 458CT17.2.4]
Antibodies
Western Blot: 1:100~1600,

Type: Primary
Antigen: GAD2
Clonality: Monoclonal
Clone: 458CT17.2.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR