Anti-IgG2a Mouse Monoclonal Antibody [clone: MRG2a-83]

Supplier: BioLegend
407501 407501 407502 407502
407502-BLEA 190.1 USD
407502-BL CA407501-BL 407501-BL CA407502-BL
Anti-IgG2a Mouse Monoclonal Antibody [clone: MRG2a-83]
Antibodies
Purified anti-rat IgG2a [MRG2a-83]; Isotype: Mouse IgG; Reactivity: Rat; Apps: ELISA, FC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA. For ELISA capture, the suggested use of this reagent is 5.0 μg per ml. It's recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgG2a
Clonality: Monoclonal
Clone: MRG2a-83
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR