Anti-CD8a Mouse Monoclonal Antibody [clone: OX-8]

Supplier: BioLegend
201701 201701
201701-BLEA 133.83 USD
CA201701-BL 201701-BL
Anti-CD8a Mouse Monoclonal Antibody [clone: OX-8]
Antibodies
Purified anti-rat CD8a [OX-8]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC, IHC, IP, WB; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: OX-8
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR