Anti-CD45RA Mouse Monoclonal Antibody [clone: OX-33]

Supplier: BioLegend
202301 202301
202301-BLEA 133.83 USD
202301-BL CA202301-BL
Anti-CD45RA Mouse Monoclonal Antibody [clone: OX-33]
Antibodies
Purified anti-rat CD45RA [OX-33]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: OX-33
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR