Anti-CD106 Mouse Monoclonal Antibody [clone: MR106]

Supplier: BioLegend
200401 200402 200402 200401
200402-BLEA 282.81 USD
200402-BL 200401-BL CA200402-BL CA200401-BL
Anti-CD106 Mouse Monoclonal Antibody [clone: MR106]
Antibodies
Purified anti-rat CD106 [MR106]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC, IHC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD106
Clonality: Monoclonal
Clone: MR106
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR