Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-REL Mouse Monoclonal Antibody [clone: 318CT41.3.2]

Supplier: Abgent
AM1937B
Western Blot: 1:120~1000,

Type: Primary
Antigen: REL
Clonality: Monoclonal
Clone: 318CT41.3.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
400 µL AM1937B 89520-994 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol REL
Antigen Name V-rel Avian Reticuloendotheliosis Viral Oncogene homolog
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 318CT41.3.2
Host Mouse
Isotype IgG2b kappa
Western Blot Yes
ELISA Yes
Antigen Synonyms c-Rel, 4.1G, 4.1-G
Gene ID 5966
Concentration 0.5mg/ml
Immunogen This REL monoclonal antibody is generated from mouse immunized with REL recombinant protein.
Molecular Weight 68.52 kDa
Purification Protein G Purification
Sequence AYNPYIEIIEQPRQRGMRFRYKCEGRSAGSIPGEHSTDNNRTYPSIQIMNYYGKGKVRITLVTKNDPYKPHPHDLVGKDCRDGYYEAEFGQERRPLFFQNLGIRCVKKKEVKEAIITRIKAGINPFNVPEKQLNDIEDCDLNVVRLCFQVFLPDEHGNLTTALPPVVSNPIYDNRAPNTAELRICRVNKNCGSVRGGDEIFLLCDKVQKDDIEVRFVLNDWEAKGIFSQADVHRQVAIVFKTPPYCKAITEPVTVKMQLRRPSDQEVSESMDFRYLPDEKDTYGNKAKKQKTTLLFQKLC
Storage Buffer PBS with 0.09% (W/V) sodium azide, pH7.4
Storage Temperature Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C in small aliquots to prevent freeze-thaw cycles
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty