Anti-Prohibitin Rabbit Polyclonal Antibody [clone: Poly6031]

Supplier: BioLegend
603102 603101
CA603102-BL CA603101-BL
Anti-Prohibitin Rabbit Polyclonal Antibody [clone: Poly6031]
Purified anti-Prohibitin [Poly6031]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human; Apps: WB; Size: 200 μl

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Prohibitin
Clonality: Polyclonal
Clone: Poly6031
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR