Anti-TLE4 Mouse Monoclonal Antibody [clone: 384CT24.1.3.4]

Supplier: Abcepta
AM2017A
89512-164EA 637.11 USD
89512-164
Anti-TLE4 Mouse Monoclonal Antibody [clone: 384CT24.1.3.4]
Antibodies
Western Blot: 1:100~3200,

Type: Primary
Antigen: TLE4
Clonality: Monoclonal
Clone: 384CT24.1.3.4
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR