Anti-p97/VCP Mouse Monoclonal Antibody [clone: 4G9]

Supplier: BioLegend
636801 636802 636802 636801
636801-BLEA 120.94 USD
636801-BL 636802-BL CA636801-BL CA636802-BL
Anti-p97/VCP Mouse Monoclonal Antibody [clone: 4G9]
Antibodies
Purified anti-p97/VCP [4G9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IF, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, suggested working dilution(s): Use 5 μg per 5 ml antibody dilution buffer for each mini-gel. For immunofluorescence microscopy: Use a dilution range of 1~4 μg/ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: p97/VCP
Clonality: Monoclonal
Clone: 4G9
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR