Anti-p63 (DN) Rabbit Polyclonal Antibody [clone: Poly6190]

Supplier: BioLegend
619001 619002 619002 619001
619002-BLEA 574.47 USD
619002-BL 619001-BL CA619002-BL CA619001-BL
Anti-p63 (DN) Rabbit Polyclonal Antibody [clone: Poly6190]
Antibodies
Purified anti-p63 (ΔN) [Poly6190]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human (recognizes δN-isoforms); Apps: WB, IHC; Size: 200 μl

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: p63 (DN)
Clonality: Polyclonal
Clone: Poly6190
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Human (recognizes δN-isoforms)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR