Anti-p63 (TA) Rabbit Polyclonal Antibody [clone: Poly6189]

Supplier: BioLegend
618901 618901 618902 618902
618901-BLEA 129.68 USD
CA618901-BL CA618902-BL 618901-BL 618902-BL
Anti-p63 (TA) Rabbit Polyclonal Antibody [clone: Poly6189]
Purified anti-p63 (TA) [Poly6189]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human, recognizes TA-isoforms; Apps: WB, IF, IHC; Size: 200 μl

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: p63 (TA)
Clonality: Polyclonal
Clone: Poly6189
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Human;recognizes TA-isoforms
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR