Anti-p38 MAPK Rabbit Polyclonal Antibody [clone: Poly6224]

Supplier: BioLegend
622402 622402 622401 622401
622401-BLEA 105 USD
622401-BL CA622402-BL CA622401-BL 622402-BL
Anti-p38 MAPK Rabbit Polyclonal Antibody [clone: Poly6224]
Purified anti-p38 MAPK [Poly6224]; Isotype: Rabbit Polyclonal IgG; Reactivity: Mouse, Rat, Human; Apps: WB, IHC; Size: 200 μl

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. For IHC, use a 1:100 dilution of antibody for staining. Antigen retrieval for IHC of formalin-fixed paraffin-embedded tissue using 0.01 M sodium citrate buffer is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: p38 MAPK
Clonality: Polyclonal
Clone: Poly6224
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR