Anti-NPM/B23 Phospho (Thr199) Rabbit Polyclonal Antibody [clone: Poly6192]

Supplier: BioLegend
619201
CA619201-BL
Anti-NPM/B23 Phospho (Thr199) Rabbit Polyclonal Antibody [clone: Poly6192]
Purified anti-NPM/B23 Phospho (Thr199) [Poly6192]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human, reacts against Thr199-phosphorylated NPM/B23; Apps: WB, IHC; Size: 50 μl

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. For IHC, use a 1:50 dilution of antibody for staining. Antigen retrieval using 0.01 M sodium citrate buffer is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: NPM/B23 Phospho (Thr199)
Clonality: Polyclonal
Clone: Poly6192
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Human;reacts against Thr199-phosphorylated NPM/B23
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR