Anti-NF-kB p65 Rabbit Polyclonal Antibody [clone: Poly6226]

Supplier: BioLegend
622602 622602 622601 622601
622601-BLEA 137.49 USD
CA622602-BL 622601-BL 622602-BL CA622601-BL
Anti-NF-kB p65 Rabbit Polyclonal Antibody [clone: Poly6226]
Antibodies
Purified anti-NF-κB p65 [Poly6226]; Isotype: Rabbit Polyclonal IgG; Reactivity: Mouse, Rat, Human; Apps: WB, IF, IHC, IP; Size: 200 μl

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.05 μg per ml. For immunofluorescence microscopy, the suggested use of the reagent is a dilution range of 1:100~1:400. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: NF-kB p65
Clonality: Polyclonal
Clone: Poly6226
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR