Anti-CD61 Armenian Hamster Monoclonal Antibody [clone: 2C9.G2 (HMbeta3-1)]

Supplier: BioLegend
104302 104302
104302-BLEA 250.71 USD
CA104302-BL 104302-BL
Anti-CD61 Armenian Hamster Monoclonal Antibody [clone: 2C9.G2 (HMbeta3-1)]
Antibodies
Purified anti-mouse/rat CD61 [2C9.G2 (HMβ3-1)]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC, IHC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD61
Clonality: Monoclonal
Clone: 2C9.G2 (HMbeta3-1)
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR