Anti-Mac-2 (Galectin-3) Mouse Monoclonal Antibody [clone: Gal397]

Supplier: BioLegend
126702 126702 126701 126701
126702-BLEA 278.53 USD
126702-BL CA126701-BL CA126702-BL 126701-BL
Anti-Mac-2 (Galectin-3) Mouse Monoclonal Antibody [clone: Gal397]
Antibodies
Purified anti-mouse/human Mac-2 (Galectin-3) [Gal397]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Mouse; Apps: WB, IF, FC, ICFC; Size: 500 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested working dilution is 0.5-1.0 μg/ml antibody dilution buffer for each mini-gel. For <a title="View Protocol" href="/media_assets/support_protocol/BioLegend_Surface_Staining_Flow_Protocol_091012.pdf" target="_blank">immunofluorescent staining</a>, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Mac-2 (Galectin-3)
Clonality: Monoclonal
Clone: Gal397
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR